AMBH303C4DBA Ambeed, Inc.

Benzyl 2-methyl-2-(methylsulfonyl)pent-4-enoate

  • MDL number MFCD30530547
  • Cas Number : 1942858-50-5
  • Molecular Weight : 282.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1942858-50-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1942858-50-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart