AMBH303C4DBD Ambeed, Inc.

Methyl 4-(chloromethyl)-3,5-difluorobenzoate

  • MDL number MFCD17676601
  • Cas Number : 1263283-69-7
  • Molecular Weight : 220.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1263283-69-7.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/1263283-69-7.html
pictograms GHS05
hstatements H314-H318
Availability details for
Quantity
  • Availability
  • Add to Cart