AMBH303C4DEE Ambeed, Inc.

2-(3-Fluoro-5-methoxyphenyl)acetonitrile

  • MDL number MFCD08741379
  • Cas Number : 914637-31-3
  • Molecular Weight : 165.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/914637-31-3.html

Safety Information

signalword Danger
pcodes P261-P280-P305+P351+P338-P311
MSDS https://www.ambeed.com/products/914637-31-3.html
pictograms GHS06
hstatements H302+H312-H315-H319-H331-H335
Availability details for
Quantity
  • Availability
  • Add to Cart