AMBH303C4E18 Ambeed, Inc.

Ethyl 5-amino-1-(pyridin-2-yl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD01124472
  • Cas Number : 69722-29-8
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/69722-29-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/69722-29-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart