AMBH303C4E1D Ambeed, Inc.

9-([1,1'-Biphenyl]-3-yl)-3-bromo-9H-carbazole

  • MDL number MFCD28138081
  • Cas Number : 1428551-28-3
  • Molecular Weight : 398.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1428551-28-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1428551-28-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart