AMBH303C4E34 Ambeed, Inc.

Ethyl 3-bromo-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate hydrochloride

  • MDL number MFCD10566311
  • Cas Number : 1170568-70-3
  • Molecular Weight : 310.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1170568-70-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1170568-70-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart