AMBH303C4E8B Ambeed, Inc.

4-(Bromomethyl)-1-chloro-2-(trifluoromethoxy)benzene

  • MDL number MFCD06660249
  • Cas Number : 886500-93-2
  • Molecular Weight : 289.4800

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/886500-93-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/886500-93-2.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart