AMBH303C4EB2 Ambeed, Inc.

3-(1H-Imidazol-5-yl)pyridine

  • MDL number MFCD10565593
  • Cas Number : 51746-85-1
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/51746-85-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/51746-85-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart