AMBH303C4EC1 Ambeed, Inc.

6-Bromo-4-nitro-2,3-dihydro-1H-inden-5-ol

  • MDL number MFCD23136024
  • Cas Number : 139515-86-9
  • Molecular Weight : 258.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/139515-86-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/139515-86-9.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart