AMBH303C4EF7 Ambeed, Inc.

4-Methylmorpholin-2-one hydrochloride

  • MDL number MFCD27920772
  • Cas Number : 1820735-05-4
  • Molecular Weight : 151.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1820735-05-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1820735-05-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart