AMBH303C4F5C Ambeed, Inc.

2,7-Dibromo-9,9-didecyl-9H-fluorene

  • MDL number MFCD00971288
  • Cas Number : 175922-78-8
  • Molecular Weight : 604.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/175922-78-8.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/175922-78-8.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart