AMBH303C4F7C Ambeed, Inc.

tert-Butyl 1-(furan-2-ylmethyl)pyrrolidine-2-carboxylate

  • MDL number MFCD23135229
  • Cas Number : 1787375-26-1
  • Molecular Weight : 251.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1787375-26-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1787375-26-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart