AMBH303C4F82 Ambeed, Inc.

Tetrabutylammonium hydroxide hydrate

  • MDL number MFCD28963519
  • Cas Number : 74296-38-1
  • Molecular Weight : 277.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 40
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/74296-38-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/74296-38-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart