AMBH303C4F85 Ambeed, Inc.

4-Fluoro-2-nitrobenzotrifluoride

  • MDL number MFCD07782061
  • Cas Number : 182289-81-2
  • Molecular Weight : 209.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/182289-81-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/182289-81-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart