AMBH303C4FA8 Ambeed, Inc.

1,4-Di(2-thienyl)-1,4-butanedione

  • MDL number MFCD00208362
  • Cas Number : 13669-05-1
  • Molecular Weight : 250.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/13669-05-1.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/13669-05-1.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart