AMBH303C4FB2 Ambeed, Inc.

4-Bromo-1-isobutyl-3-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD24448782
  • Cas Number : 1437794-62-1
  • Molecular Weight : 271.0800

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1437794-62-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1437794-62-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart