AMBH303C4FC0 Ambeed, Inc.

Methyl 4-chloro-6-methoxypicolinate

  • MDL number MFCD18257705
  • Cas Number : 204378-37-0
  • Molecular Weight : 201.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/204378-37-0.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/204378-37-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart