AMBH303C5000 Ambeed, Inc.

5-Ethynylpyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD00871973
  • Cas Number : 59989-18-3
  • Molecular Weight : 136.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/59989-18-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/59989-18-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart