AMBH303C5016 Ambeed, Inc.

(4-((4-(Ethylamino)-5-(trifluoromethyl)pyrimidin-2-yl)amino)-2-fluoro-5-methoxyphenyl)(morpholino)methanone

  • MDL number MFCD25977005
  • Cas Number : 1351761-44-8
  • Molecular Weight : 443.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1351761-44-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1351761-44-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart