AMBH303C5019 Ambeed, Inc.

XL388

  • MDL number MFCD24386875
  • Cas Number : 1251156-08-7
  • Molecular Weight : 455.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (7-(6-Aminopyridin-3-yl)-2,3-dihydrobenzo[f][1,4]oxazepin-4(5H)-yl)(3-fluoro-2-methyl-4-(methylsulfonyl)phenyl)methanone
Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1251156-08-7.html

Safety Information

signalword Warning
pcodes P280-P304+P340+P312-P305+P351+P338-P337+P313
MSDS https://www.ambeed.com/products/1251156-08-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart