AMBH303C5029 Ambeed, Inc.

5-Bromo-2-iodobenzonitrile

  • MDL number MFCD00672950
  • Cas Number : 121554-10-7
  • Molecular Weight : 307.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-122
Purity 95
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 310.4�t 760 mmHg
Product Link https://www.ambeed.com/products/121554-10-7.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/121554-10-7.html
pictograms GHS07
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart