AMBH303C5072 Ambeed, Inc.

5-(3,4-Dichlorophenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD01191258
  • Cas Number : 28004-64-0
  • Molecular Weight : 246.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 209.3
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 422.5�t 760 mmHg
Product Link https://www.ambeed.com/products/28004-64-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/28004-64-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart