AMBH303C507A Ambeed, Inc.

4-(1-Aminoethyl)aniline

  • MDL number MFCD07371388
  • Cas Number : 90434-58-5
  • Molecular Weight : 136.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/90434-58-5.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/90434-58-5.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart