AMBH303C5080 Ambeed, Inc.

4-Methylesculetin

  • MDL number MFCD00006859
  • Cas Number : 529-84-0
  • Molecular Weight : 192.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/529-84-0.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501A
MSDS https://www.ambeed.com/products/529-84-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart