AMBH303C5091 Ambeed, Inc.

2,4-Bipyridine

  • MDL number MFCD00006217
  • Cas Number : 581-47-5
  • Molecular Weight : 156.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/581-47-5.html

Safety Information

signalword Warning
pcodes P261-P264-P270-P271-P280-P301+P312+P330-P302+P352+P312+P362+P364-P304+P340+P312-P305+P351+P338+P337+P313-P501
MSDS https://www.ambeed.com/products/581-47-5.html
pictograms GHS07
hstatements H302+H312+H332-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart