AMBH303C50E9 Ambeed, Inc.

Diethyl 2,2'-((3-cyano-4-(2-ethoxy-2-oxoethyl)-5-(ethoxycarbonyl)thiophen-2-yl)azanediyl)diacetate

  • MDL number MFCD09839706
  • Cas Number : 58194-26-6
  • Molecular Weight : 454.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/58194-26-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/58194-26-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart