AMBH303C5106 Ambeed, Inc.

2,6-Diiodopyridine

  • MDL number MFCD07366728
  • Cas Number : 53710-17-1
  • Molecular Weight : 330.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-180
Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 316.8�t 760 mmHg
Product Link https://www.ambeed.com/products/53710-17-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/53710-17-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart