AMBH303C5116 Ambeed, Inc.

Sodium diethylcarbamodithioate

  • MDL number MFCD00041028
  • Cas Number : 148-18-5
  • Molecular Weight : 171.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95�t N/A mmHg
Synonym (E)-1-(6-Methoxynaphthalen-2-yl)-3-(pyridin-3-yl)prop-2-en-1-one
Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 176.4�t 760 mmHg
Product Link https://www.ambeed.com/products/148-18-5.html

Safety Information

signalword Warning
pcodes P233-P260-P261-P264-P271-P280-P302+P352-P304-P304+P340-P305+P351+P338-P312-P321-P332+P313-P337+P313-P340-P362-P403-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/148-18-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart