AMBH303C5127 Ambeed, Inc.

2-(Chloromethyl)-3-fluoropyridine

  • MDL number MFCD11520697
  • Cas Number : 149489-32-7
  • Molecular Weight : 145.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/149489-32-7.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/149489-32-7.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart