AMBH303C5133 Ambeed, Inc.

N-(3-Nitrophenyl)propionamide

  • MDL number MFCD00791259
  • Cas Number : 7470-50-0
  • Molecular Weight : 194.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-104
Purity 97
Flash Point 186.6
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 384.9�t 760 mmHg
Product Link https://www.ambeed.com/products/7470-50-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/7470-50-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart