AMBH303C5144 Ambeed, Inc.

UNC669

  • MDL number MFCD26936345
  • Cas Number : 1314241-44-5
  • Molecular Weight : 338.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (5-Bromopyridin-3-yl)(4-(pyrrolidin-1-yl)piperidin-1-yl)methanone
Purity 98
Flash Point 230.8
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 458.0�0�t 760 mmHg
Product Link https://www.ambeed.com/products/1314241-44-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1314241-44-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart