AMBH303C5148 Ambeed, Inc.

3-(3-Methoxyphenyl)pyrrolidine oxalate(2:1)

  • MDL number MFCD28016703
  • Cas Number : 1610028-40-4
  • Molecular Weight : 444.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1610028-40-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1610028-40-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart