AMBH303C515D Ambeed, Inc.

AZD8055

  • MDL number MFCD16660191
  • Cas Number : 1009298-09-2
  • Molecular Weight : 465.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (5-(2,4-Bis((S)-3-methylmorpholino)pyrido[2,3-d]pyrimidin-7-yl)-2-methoxyphenyl)methanol
Purity 98
Flash Point 373.7
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 694.3�0�t 760 mmHg
Product Link https://www.ambeed.com/products/1009298-09-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1009298-09-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart