AMBH303C5175 Ambeed, Inc.

5-(tert-Butyl)-1H-indole

  • MDL number MFCD20668007
  • Cas Number : 92462-70-9
  • Molecular Weight : 173.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/92462-70-9.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/92462-70-9.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart