AMBH303C518A Ambeed, Inc.

1-(Benzyloxy)-2-methoxy-4-(prop-1-en-1-yl)benzene

  • MDL number MFCD00026985
  • Cas Number : 120-11-6
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-63
Purity 98
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 388.638�t 760 mmHg
Product Link https://www.ambeed.com/products/120-11-6.html

Safety Information

signalword Danger
pcodes P261-P273-P280-P305+P351+P338
hcodes 9
MSDS https://www.ambeed.com/products/120-11-6.html
pictograms GHS05,GHS07,GHS09
hstatements H315-H317-H318-H335-H410
Availability details for
Quantity
  • Availability
  • Add to Cart