AMBH303C51CD Ambeed, Inc.

Methyl 2-(2-oxo-1,2,3,4-tetrahydroquinolin-3-yl)acetate

  • MDL number MFCD24553699
  • Cas Number : 61164-72-5
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150.5-151
Purity 97
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 387.8�0�t 760 mmHg
Product Link https://www.ambeed.com/products/61164-72-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/61164-72-5.html
pictograms GHS07
hstatements H302-H315-H317-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart