AMBH303C51EF Ambeed, Inc.

6-Bromo-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride

  • MDL number MFCD28986166
  • Cas Number : 1810069-91-0
  • Molecular Weight : 262.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1810069-91-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1810069-91-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart