AMBH303C51FA Ambeed, Inc.

Montelukast sodium

  • MDL number MFCD00931431
  • Cas Number : 151767-02-1
  • Molecular Weight : 608.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Sodium (R,E)-2-(1-(((1-(3-(2-(7-chloroquinolin-2-yl)vinyl)phenyl)-3-(2-(2-hydroxypropan-2-yl)phenyl)propyl)thio)methyl)cyclopropyl)acetate
Purity 98
Flash Point 407.7
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/151767-02-1.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/151767-02-1.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart