AMBH303C5232 Ambeed, Inc.

Boc-D-Glu(Ome)-OH.DCHA

  • MDL number MFCD01861340
  • Cas Number : 76379-02-7
  • Molecular Weight : 442.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Dicyclohexylamine (R)-2-((tert-butoxycarbonyl)amino)-5-methoxy-5-oxopentanoate
Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/76379-02-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/76379-02-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart