AMBH303C5233 Ambeed, Inc.

endo-8-Azabicyclo[3.2.1]octan-3-yl 2-hydroxy-2,2-diphenylacetate hydrochloride

  • MDL number MFCD13194790
  • Cas Number : 63516-30-3
  • Molecular Weight : 373.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1R,3r,5S)-8-Azabicyclo[3.2.1]octan-3-yl 2-hydroxy-2,2-diphenylacetate hydrochloride
Purity 95
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/63516-30-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/63516-30-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart