AMBH303C5240 Ambeed, Inc.

(R)-1-(3-Chloro-4-fluorophenyl)ethanamine hydrochloride

  • MDL number MFCD12758039
  • Cas Number : 1257106-65-2
  • Molecular Weight : 210.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1257106-65-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/1257106-65-2.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart