AMBH303C5241 Ambeed, Inc.

[4-(1,3-Thiazol-2-yl)phenyl]methylamine

  • MDL number MFCD09025834
  • Cas Number : 672324-87-7
  • Molecular Weight : 190.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/672324-87-7.html

Safety Information

signalword Danger
pcodes P261-P280-P301+P310-P305+P351+P338
hcodes 8
MSDS https://www.ambeed.com/products/672324-87-7.html
pictograms GHS05,GHS06
hstatements H301-H315-H318-H335
Availability details for
Quantity
  • Availability
  • Add to Cart