AMBH303C524C Ambeed, Inc.

1-(2-Chlorophenyl)ethanamine hydrochloride

  • MDL number MFCD11841333
  • Cas Number : 856629-37-3
  • Molecular Weight : 192.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/856629-37-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/856629-37-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart