AMBH303C525D Ambeed, Inc.

4-Amino-6-chloro-5-methylpyrimidine

  • MDL number MFCD09991585
  • Cas Number : 14394-56-0
  • Molecular Weight : 143.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237-238°C
Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 292.4°C at 760 mmHg
Product Link https://www.ambeed.com/products/14394-56-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/14394-56-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart