AMBH303C5270 Ambeed, Inc.

2-(4-(2-(Trifluoromethyl)benzoyl)piperazin-1-yl)thiazole-5-carboxamide

  • MDL number MFCD13193183
  • Cas Number : 916888-66-9
  • Molecular Weight : 384.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/916888-66-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/916888-66-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart