AMBH303C528E Ambeed, Inc.

Ytterbium(III) chloride hexahydrate

  • MDL number MFCD00149885
  • Cas Number : 10035-01-5
  • Molecular Weight : 326.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 865
Synonym N/A
Purity 99.9
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/10035-01-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/10035-01-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart