AMBH303C52A4 Ambeed, Inc.

2-(1'-(tert-Butoxycarbonyl)spiro[chroman-2,4'-piperidine]-4-yl)acetic acid

  • MDL number MFCD12198637
  • Cas Number : 952681-82-2
  • Molecular Weight : 361.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/952681-82-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/952681-82-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart