AMBH303C52E5 Ambeed, Inc.

tert-Butyl spiro[indoline-3,4'-piperidine]-1-carboxylate hydrochloride

  • MDL number MFCD15071705
  • Cas Number : 1243474-66-9
  • Molecular Weight : 324.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1243474-66-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1243474-66-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart