AMBH303C52E8 Ambeed, Inc.

Trifluoro(pyrrolidin-1-ium-1-ylmethyl)borate

  • MDL number MFCD20134172
  • Cas Number : 1686150-29-7
  • Molecular Weight : 152.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1686150-29-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1686150-29-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart