AMBH303C530E Ambeed, Inc.

6,7-Dimethoxy-2-methyl-1-(3,4,5-trimethoxybenzyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD30536255
  • Cas Number : 2048273-58-9
  • Molecular Weight : 423.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/2048273-58-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2048273-58-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart